missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Beatus Bottle Top Dispenser

Robust and reliable dispensing; Fisherbrand™ Beatus Bottle Top Dispenser has re-circulation valve for re-directing the liquid into the bottle for bubble free dispensing without any loss of reagent.

236.00 € - 692.00 €

Specifications

Product Type Bottletop Dispenser
Autoclavable Autoclavable (121°C, 15psi for 10-15 min.)
Certifications/Compliance ISO 8655 standards
Material Borosilicate Glass
View More Specs

Products 6
Product Code Brand Model Volume (Metric) Accuracy Increments Price Quantity & Availability  
Product Code Brand Model Volume (Metric) Accuracy Increments Price Quantity & Availability  
15518901
View accessory products View Documents
Fisherbrand™
BEAT2.5
BEAT-2.5 0.25 to 2.5 mL 0.6% (+/-0.015) 0.05 mL
236.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15538901
View accessory products View Documents
Fisherbrand™
BEAT5.0
BEAT-5 0.5 to 5 mL 0.6% (+/-0.030) 0.1 mL
498.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15528901
View accessory products View Documents
Fisherbrand™
BEAT30
BEAT-30 2.5 to 30 mL 0.6% (+/-0.180) 0.5 mL
554.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15548901
View accessory products View Documents
Fisherbrand™
BEAT60
BEAT-60 5 to 60 mL 0.6% (+/-0.0360) 1.0 mL
287.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15598891
View accessory products View Documents
Fisherbrand™
BEAT10
BEAT-10 1 to 10 mL 0.6% (+/-0.060) 0.2 mL
482.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15508901
View accessory products View Documents
Fisherbrand™
BEAT100
BEAT-100 10 to 100 mL 0.6% (+/-0.600) 2.0 mL
692.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • With re-circulation valve
  • Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
  • Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
  • Specially designed adaptor allows 360° rotation providing full flexibility
  • Springless, PTFE valve manifold ensures high chemical compatibility and jamming free valve function
  • PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
  • Adaptors to fit most of the laboratory reagent bottles
  • Calibration tool provided for convenient and quick in-lab user re-calibration


Specifications:
  • Piston: PTFE and ETP
  • Valve housing: PFA
  • Re-Circulation valve housing: PFA
  • Valve assembly: Borosilicate glass and seat
  • Discharge assembly: PTFE
  • Delivery and Inlet tube: FEP
  • Compatibility: With all reagents except HF
Specifications

Specifications

Bottletop Dispenser
Autoclavable (121°C, 15psi for 10-15 min.)
ISO 8655 standards
Borosilicate Glass
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.