missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beaucage reagent, MedChemExpress
MedChemExpress Beaucage reagent is found to be potent in causing DNA cleavage.
Brand: Medchem Express HY-100951
This item is not returnable.
View return policy
Description
Beaucage reagent is found to be potent in causing DNA cleavage.Specifications
Beaucage reagent | |
White | |
C7H4O3S2 | |
DMSO : ≥ 100 mg/mL (499.43 mM) ∣H2O : 0.14 mg/mL (0.70 mM; Need ultrasonic) | |
200.23 | |
98.0% | |
Solid |
66304-01-6 | |
Research | |
1 g | |
O=C(C1=CC=CC=C12)SS2(=O)=O | |
200.23 | |
Research | |
Cancer-programmed cell death |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only
Spot an opportunity for improvement?Share a Content Correction