missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
Brand: Bel-Art™ F13164-1923
338.67 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
- Bright red polypropylene bags include black biohazard symbol
- Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
- Sterilization patch darkens when subjected to steam sterilization
- High temperature steam autoclavable to 135°C (275°F)
- Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
- Made in USA
Specifications
Biohazard Disposal Bag | |
Autoclavable | |
26 L | |
Zip Tie | |
Single-use | |
Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish | |
Non-sterile | |
1.50 mil | |
23 in. | |
19 in. | |
23 in. |
19W x 23H in. (48 x 58cm) | |
5 to 9 gal. | |
ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact | |
Red | |
Used for biohazard waste | |
Polypropylene | |
0.279 mm | |
19 x 23 in. | |
58 cm | |
48 cm | |
58 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil > 19W x 23H in. (48 x 58cm); 6-9 gal.
Spot an opportunity for improvement?Share a Content Correction