missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Acrylic Desiccator Cabinet

Used in refrigerators or at room temperature.

792.92 € - 909.65 €

Specifications

Material Acrylic
Door Style Gasketed
For Use With (Application) Used in refrigerators/at room temperature for light-sensitive materials
Holds 32 Cans
Includes Tray-like 3 shelves with perforated bottoms and slide in/out on pre-set rails, stainless steel twist locking latch for door
View More Specs

Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
12693496
View Documents
Bel-Art™
F42066-0000
Clear
792.92 €
Each
Estimated Shipment: 06-03-2025
to see stock.
Add to basket
 
15244939
View Documents
Bel-Art™
F42066-0001
Bronze
909.65 €
Each
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
 
Description

Description

  • Acrylic cabinet with removable, tray-like shelves with perforated bottoms and slide in/out on pre-set rails
  • Door opens from right to left and secured with stainless steel latch
  • Available with three shelves
  • Comes in clear color and bronze color for light-sensitive materials
Specifications

Specifications

Acrylic
Used in refrigerators/at room temperature for light-sensitive materials
Tray-like 3 shelves with perforated bottoms and slide in/out on pre-set rails, stainless steel twist locking latch for door
0.82 cu. ft.
12 in.
12 in.
12 in.
Gasketed
32 Cans
3 removable tray-like shelves
12 x 12 in. (30.4 x 30.4 cm)
30.4 cm
30.4 cm
30.4 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.