missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Autoradiography Pens

Easy marking of film with nontoxic and nonradioactive phosphorescent material

106.00 € - 111.00 €

Specifications

Product Type Non Radioactive Marker
Color Bright Green
Includes Pocket clip and screw-knob advance to adjust writing tip
Quantity Each
Length (English) 4.5 in.
View More Specs

Includes: Pocket clip and screw-knob advance to adjust writing tip.

Products 2
Product Code Brand For Use With (Application) Price Quantity & Availability  
Product Code Brand For Use With (Application) Price Quantity & Availability  
12320469
View Documents
Bel-Art™
F13350-0000
Ideal for marking various films that are to be X-rayed with serial numbers, dates and exposure periods, film position markings This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10719475
View Documents
Bel-Art™
F13351-0000
Ideal for marking radiography films like the Kodak XLS This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Easy marking of film with nontoxic and nonradioactive phosphorescent material

  • Solid phosphorescent material will not leak or dry out, (no bladder, vial or liquids to spill) Nonradioactive energy may be adjusted by exposure to white incandescent or fluorescent light
  • 3 to 4 minute energy emission leaves distinct impression without overexposing film
  • Bright green markings do not glow in the dark but are visible under normal conditions and on X-ray film
  • No Beta or other isotope emissions; Spectral peak, 5600Å
  • Screw knob advances writing tip; Pens are 4 1/2 in. long (11.4cm)
  • Not appropriate for phosphor screens

High Energy Level Markers
Greater energy emission strength and slightly longer energy emission duration
Suitable for marking X-Ray films similar to Kodak™ XLS
No Beta or other isotope emissions
Marking is unaffected by chilled films and an intensifier screen will not affect intensity
Not appropriate for phosphor screens

Specifications

Specifications

Non Radioactive Marker
Pocket clip and screw-knob advance to adjust writing tip
4.5 in.
Bright Green
Each
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.