missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Bacterial Cell Spreaders
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
Brand: Bel-Art™ F37736-0009
This item is not returnable.
View return policy
Description
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
- Autoclavable and may be flame sterilized
- Features triangular end with smooth rounded surfaces throughout
- Smaller 16mm width bar suitable for use with 6cm plates
- Larger 25mm width bar suitable for use with 9cm plates
- Use with Inoculating Turntable (Mfr. No.F40913-0000)
- Both sizes 30cm long
Specifications
Autoclavable | |
12 in. | |
Stainless Steel | |
Sterile |
Inoculating turntable, 9cm inoculating plates | |
30.2 cm | |
Bacterial Cell Spreader | |
Easily perform inoculation of petri dishes |