Learn More
Bel-Art™ SP Scienceware™ Bag Twist Tie Cord
Strong twist tie bag closures easier to use than string or rubber bands
Brand:  Bel-Art™ H13190-0000
100.74 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 11707035
  • 129.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Cut to optimum length and twist together for best closure performance
  • Use to bundle hard-to-handle items like rods, wires or tubes
  • Made of soft, copper-tinned, annealed iron wire sealed in extruded virgin vinyl
  • Autoclavable 121°C for 30 minutes
  • 1500 ft. per roll
Specifications
Specifications
1500 ft
Use to close bags or to package rods, wires and tubes