Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.)
Brand:  Bel-Art™ H19559-0000
 View more versions of this product
Product Code. 11744935
  • 69.75 € / Pack of 12
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Specifications
Barbed Fitting
Polypropylene
For 3/16 in. to 1/2 in. tubing (4.7 to 9.5mm)
Clear
Autoclavable
4.7 to 9.5mm
Reducing connector