missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Sterileware™ Bent Handle Scoop
Features an ergonomic, curved handle design to elevate the hand from the sample, reducing risk of contamination
42.90 € - 43.15 €
Specifications
Product Type | Scoop |
---|---|
Material | Polystyrene |
Autoclavable | Not Autoclavable |
Bottom Type | Flat |
Handle Type | Bent, Curved |
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
13452489
|
Bel-Art™
H36902-2006 |
60 mL | 7.62 x 1.62 x 1.12cm (19.4 x 4.1 x 2.9 cm) |
43.15 €
Pack of 10 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
||||
13472489
|
Bel-Art™
H36904-2006 |
125 mL | 7.68 x 2 x 1.31 in. (19.4 x 5.1 x 3.3 cm) | N/A | |||||
Description
- Curved handle helps prevent incidental contact with sample source material and exposure to hazardous substances
- Flat bottom keeps scoop upright when placed on a flat surface; can be used as a weigh boat
- Made from high-impact, lightweight, FDA grade polystyrene
- Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Sterilization
- Individually packaged and ready for one time use and disposal
Specifications
Scoop | |
Not Autoclavable | |
Bent, Curved | |
Sterile | |
Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances | |
19.4 cm |
Polystyrene | |
Flat | |
Single-use | |
FDA | |
7.62 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title