missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Buchner Table-Top Funnels, 18 in. (45.7cm) I.D.

Filter large volumes quickly with a polyethylene plastic Table-Top Buchner Funnel.

486.00 € - 1190.00 €

Specifications

Material Polyethylene
View More Specs

Products 3
Product Code Brand Description Diameter (Metric)Top Height (Metric) Price Quantity & Availability  
Product Code Brand Description Diameter (Metric)Top Height (Metric) Price Quantity & Availability  
12997300
missing translation for 'viewDocuments'
Bel-Art™
H14626-3518
Funnel with Medium Porosity Removable Plate (18 in) 45.7 cm 285.75 mm
1190.00 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
10369009
missing translation for 'viewDocuments'
Bel-Art™
H14628-0000
Funnel with Perforated Fixed Plate (18 in) 45.7 cm 285.75 mm
1015.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
15199072
missing translation for 'viewDocuments'
Bel-Art™
H14625-3510
Funnel with Medium Porosity Fixed Plate (10.25 in) 26 cm 20.3 cm
486.00 €
Each
Estimated Shipment: 28-03-2025
to see stock.
Add to basket
 
Description

Description

  • Removable hose barbed connector accepts 12.7mm (1/2 in.) I.D. tube and threads into 3/4 in. NPT bung on the funnel
  • Fixed or removable Fritware™ Porous Filter Plates supported by a multiple ring grid; HDPE, 6.4mm (1/4 in.) thick, with a non-porous ring at the periphery of the plate which seals filter paper when used
  • Fritware Porous Filter Plates available in two porosities (medium and coarse); medium is 45-90 microns while the coarse is 90-130 microns; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF)
  • Fixed or removable Perforated Filter Plates are made of 4.8mm (3/16 in.) thick HDPE with 4.8mm (3/16 in.) perforations on 11mm (7/16 in.) centers
  • Perforated Filter Plates available in two versions (fixed or removable); fixed for coarse filtration or use with a cloth or paper filter; removable for use in precious metals recovery; use temperature range maximum 52ºC (125ºF), minimum -20ºC (-4ºF)
  • Rim to Plate: 8 in. (20.3 cm)
  • Use the Funnel Trowel (F36820-0118) to quickly and efficiently smooth fissures appearing during the filtration process
Specifications

Specifications

Polyethylene
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.