missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ ProCulture Cell Lifter
Economical and reusable tools to scrape cell lysates or remove and harvest tissue cultures from all types of dishes
Brand: Bel-Art™ F37756-0010
This item is not returnable.
View return policy
Description
Economical and reusable tools to scrape cell lysates or remove and harvest tissue cultures from all types of dishes
- Narrow blade 0.6 in. (15mm)
- Suitable for use in 35 and 60mm Petri dishes and 6- and 12-well cell culture clusters
- Angled and chiseled blade with smooth handle molded from polypropylene for good chemical resistance
- Reusable
- Autoclavable at 121°C (250°F)
- 7.75 × 0.5 in. (19.7 × 1.3cm)
Specifications
Polypropylene | |
Scraping up cell lysates and harvesting tissue cultures from various cell culture dishes | |
Reusable | |
21 cm | |
0.625 in. |
Autoclavable | |
35mm and 60mm petri dishes and 6- and 12-well cell culture clusters | |
8.25 in. | |
1.6 cm | |
Cell Lifter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction