missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Clavies™ Biohazard Bag Holders
Keep bags open and ready to use
86.45 € - 102.00 €
Specifications
Product Type | Biohazard Bag Holder |
---|---|
Autoclavable | Autoclavable |
Color | Silver, Orange |
Disposable | Reusable |
For Use With (Application) | Designed to securely support the bag and its contents |
Product Code | Brand | Height (English) | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Height (Metric) | Price | Quantity & Availability | |||||
11797864
|
Bel-Art™
F13192-0003 |
27.2 in. | 69.6 cm |
102.00 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
11322154
|
Bel-Art™
F13192-0001 |
18.5 in. | 47 cm |
86.45 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
11787864
|
Bel-Art™
F13192-0002 |
21.5 in. | 54.6 cm |
92.95 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
Description
- Economical, lightweight stainless-steel wire frame
- Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
- Tray serves as containment well in case of bag leakage
- Autoclavable at 121°C (250°F) and dishwasher safe
- Assembly is easy and permanent
- Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation
Specifications
Biohazard Bag Holder | |
Silver, Orange | |
Designed to securely support the bag and its contents | |
Sterile |
Autoclavable | |
Reusable | |
Stainless Steel | |
Benchtop |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ Clavies™ Biohazard Bag Holders