missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Clear View Fume Hood

Safely vent noxious or potentially hazardous fumes and dust with the SP Scienceware™ Clear View Fume Hood.

3030.00 € - 3059.86 €

Specifications

Product Type Fume Hood
Material Polycarbonate
Depth (English) 24 in.
Depth (Metric) 61 cm
Height (English) 26 in.
View More Specs

Includes: Laboratory, classroom or industrial

Products 2
Product Code Brand Width (English) Width (Metric) Price Quantity & Availability  
Product Code Brand Width (English) Width (Metric) Price Quantity & Availability  
15564034
View Documents
Bel-Art™
F50020-1010
25.87 in. 65.7 cm
3030.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17207719
View Documents
Bel-Art™
F50020-2010
43.87 in. 111.4 cm
3059.86 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Versatile 3-position sash
  • Clear polycarbonate provides optimum visibility and durability

500201010

  • Top sitting: 6 in. O.D. vent
  • O.D.: 24D x 25.8W x 26.12 in. H (61 x 65.7 x 66.4cm)
  • I.D.: 20D x 24W x 24 in. H (50.8 x 61 x 61cm)
  • Internal Space: 11.67 cu. ft. (0.18m)
500202010
  • Two Top sitting: 6 in. O.D. vent
  • O.D.: 24D x 43.8W x 26.1 in. H (61 x 111.4 x 66.4cm)
  • I.D.: 20D x 42W x 24 in. H (50.8 x 106.7 x 61cm)
  • Internal Space: 11.67 cu. ft. (0.18m)

Specifications

Specifications

Fume Hood
24 in.
26 in.
Polycarbonate
61 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.