Learn More
Bel-Art™ Cloning Cylinders
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
87.85 € - 151.00 €
Specifications
Sterility Sterile
Material Polystyrene
Product Type Cloning Cylinder
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10246182
View Documents
Bel-Art™
F37847-0300
Large
114.27 €
Pack of 50
In Stock
to see stock.
Add to basket
11795176
View Documents
Bel-Art™
H37847-0000
Small, Medium, Large (Variety)
151.00 €
Pack of 40
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11717085
View Documents
Bel-Art™
F37847-0100
Small
87.85 €
Pack of 50
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
10552683
View Documents
Bel-Art™
F37847-0200
Medium
93.40 €
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
Description
Description
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
  • Tapered
  • Good sealing surface
  • Polystyrene
  • Sterile
Order Info
Individual sizes supplied five bags of 10 cylinders each; Variety Pack contains four bags with 10 of each size
Specifications
Specifications
Sterile
Polystyrene
Cloning Cylinder