Learn More
Bel-Art™ SP Scienceware™ Complete Fittings Assortment
Reduces time spent searching for the right tubing connector or stopper
Brand:  Bel-Art™ F19625-0000
Product Code. 12319548
  • 259.00 € / Pack of 72
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This generous assortment of 72 plastic fittings has everything needed for routine laboratory set-ups.
  • Clear, hinge-top, polystyrene storage box makes it easy to find what you need
  • Includes commonly used sizes of connector and stopper styles
  • FDA grade, autoclavable polypropylene connectors: 15 Straight Tubing Connectors, 11 Stepped Tubing Connectors, 6 “T” Shaped Tubing Connectors, 10 Wye (Y) Tubing Connectors
  • Polyethylene connectors: 3 pair Quick Disconnects
  • Polyethylene stoppers: 24 Cup-Type
Specifications
Specifications
Fittings Assortment
Assortment of 72 plastic fittings