missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Cover Glass Forceps

Tips shaped to easily lift cover glass. Bel-Art™ Cover Glass Forceps are fully coated with brown Teflon™ FEP and comply with FDA regulations concerning direct food contact.

Brand:  Bel-Art™ H37942-0000

 View more versions of this product

Product Code. 10717165

  • 69.60 € / Pack of 2
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Brown PTFE FEP coating guards against scratches
  • Wide, bent tip style is perfect for lifting cover glasses
Specifications

Specifications

Bent
Teflon
Forceps
Standard
4 in.
102 mm
Cover Glass Forceps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Cover Glass Forceps > Glass Forceps, Bent 102mm long

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.