missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Cryo-Safe™ Microcentrifuge Tube and Cryogenic Vial Coolers

Benchtop-sized coolers for microcentrifuge tubes or cryogenic vials.

Brand:  Bel-Art™ F18846-2000

 View more versions of this product

Product Code. 11794124

  • 280.67 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with microcentrifuge tubes or cryogenic vials.

Holds Thirty two 0.5 or 1.5mL centrifuge tube in separate wells below 0°C for 3 Hours.

  • Body of cooler filled with a nontoxic gel that does not contact the contents of the cooler
  • Excellent for safe transport of temperature sensitive tubes
  • Polycarbonate body and lid is easy to clean
  • Stackable design with rubber feet to prevent sliding
  • Mini Coolers feature molded-in side handles, Junior and Maxi Coolers have fold down wire carrying handle that helps retain lid
  • Store -15°C coolers at -15°C for 24 hours prior to use
  • Store 0°C coolers at -20°C for a few hours prior to use
  • Store -20°C coolers at -20°C for 24 hours prior to use

 

Specifications

Specifications

14.6 cm
24.3 cm
15.7 cm
9.5 x 3.19 x 5.7 in. (24.3 x 15.7 x 14.6 cm)
2 kg
Maxi Cooler
32 (0.5 or 1.5mL centrifuge tube in separate wells below 0°C for 3 Hours)
5.7 in.
9.5 in.
3.19 in.
4.40 lb.
Polycarbonate
Gray, Red
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers