Learn More
Bel-Art™ SP Scienceware™ Delrin™ Spring Tubing Clamps
Self-closing spring operated tubing clamp is suitable for all flexible tubing
45.20 € - 64.75 €
Specifications
Type Spring Style Tubing Clamp
Material Acetal Plastic, Stainless Steel
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10294441
View Documents
Bel-Art™
H18214-0000
All flexible .46in. (1.16cm) O.D. tubing
64.75 €
Pack of 3
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
10458222
View Documents
Bel-Art™
H18213-0000
All flexible .31 in. (.78cm) O.D. tubing
45.20 €
Pack of 3
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
Description
Description
  • Positive-actionclamp opens by squeezing handles together
  • Naturally clamps shut to stop flow
  • Made from chemical- and stain-resistant Delrin™ plastic and stainless steel
Specifications
Specifications
Spring Style Tubing Clamp
Acetal Plastic, Stainless Steel