missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Dispensing Jug Rack

Rugged, corrosion-resistant polyethylene rack has 5 shelves for stacking four dispensing jugs and a spare jug or other equipment

Brand:  Bel-Art™ H11851-0000

Product Code. 12310469

  • 311.00 € / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Each shelf is 26cm (10.5 in.) high
  • Supports 91kg (200 lb.) per rack, or 18kg (40 lb.) per shelf
  • Shipped flat, ready for easy assembly
  • Shipping weight 4.7kg (10.3 lb.)
  • Bottles and jugs not included

Order Info

Compatiable with Mfr. No. H11850 dispensing jugs, not included.

Specifications

Specifications

Dispensing Jug Rack
15.25 x 13.87 x 60 in. (39 x 35 x 152cm)
15.25 in.
13.87 in.
60 in.
Not Autoclavable
39 cm
35 cm
152 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers