Learn More
Bel-Art™ SP Scienceware™ Egg-Shaped Spinbar™ Magnetic Stir Bars
Designed for use in round-bottom vessels
11.80 € - 36.30 €
Specifications
Autoclavable Autoclavable
Color White
Quantity 1
Shape Egg
View More Specs
Products 7
Product Code Brand Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity & Availability  
12358229
View Documents Promotion Details
Bel-Art™
F37130-0034
0.37 in. 9.5 mm 0.75 in. 19 mm
13.75 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
11316954
View Documents Promotion Details
Bel-Art™
F37130-0038
0.18 in. 4.7 mm 0.37 in. 9.5 mm
11.80 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
12398229
View Documents Promotion Details
Bel-Art™
F37130-0002
0.75 in. 19 mm 2 in. 50.8 mm
26.60 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
12368229
View Documents Promotion Details
Bel-Art™
F37130-0001
0.5 in. 12.7 mm 1 in. 25.4 mm
13.50 €
Each
In Stock
to see stock.
Edge
Add to basket
12378229
View Documents Promotion Details
Bel-Art™
F37130-0114
0.62 in. 16 mm 1.25 in. 32 mm
16.27 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
12388229
View Documents Promotion Details
Bel-Art™
F37130-0112
0.62 in. 16 mm 1.5 in. 38 mm
17.85 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
12308239
View Documents Promotion Details
Bel-Art™
F37130-0003
0.75 in. 19.1 mm 2.99 in. 76 mm
36.30 €
Each
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
Description
Description
  • Provide their own pivot point without a separate ring or abrupt change in contour for unrestricted spin
  • Minimal contact when used in plastic containers
  • Magnet itself is egg-shaped encapsulated in FDA grade, USP Class VI Teflon™ PTFE, except three smallest sizes have a cylindrical magnet in egg-shaped Teflon PTFE
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification
  • Autoclave safe
Specifications
Specifications
Autoclavable
White
1
Egg