missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Flowmi™ Cell Strainers for 1000μL Pipette Tips

Product Code. p-4686138 Shop All SP Scienceware Products
Click to view available options
Mesh Size:
40 μm
70 μm
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 15342931

Brand: Bel-Art™ H136800040

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Provides fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis (patent pending). Available in two porosity sizes.

  • Provide fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis
  • Preserves volume when filtering small sample volumes
  • Removes cell aggregates and creates uniform single cell suspensions
  • Decreases the potential for clogging of FLOW or FACS instruments by effectively filtering cellular debris
  • Compact tray holds 50 Flowmi Cell Strainers ready for press and fit attachment to tip; features a sliding cover for easy, one-handed use
  • Packaged in a re-sealable bag to maintain sterility when used in laminar flow hood or biohazard hood
  • Sterile, 40 μm or 70 μm porosity

Specifications

Mesh Material Polyethylene
Color White
For Use With (Application) Provide fast and efficient filtering of small volume samples (up to 1000μL) prior to FLOW or FACS™ analysis
Includes Compact tray
Product Type Cell Strainer
Sterility Sterile
For Use With (Equipment) Pipette Tips, 1000 μL, including Fisherbrand Sure-One, VWR, Axygen, Nichiryo, Eppendorf and more
Mesh Size 40 μm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.