Learn More
Bel-Art™ SP Scienceware™ Fluorescent Ruler
Useful for all fluorescent viewing and fluorescent photographic work
Brand:  Bel-Art™ F13355-0001
22.21 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 10528731
  • 24.32 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The scale will glow visibly when illuminated with ultraviolet light
  • This Metric/English scale is printed on plastic with a pink phosphorescent ink
  • The scale is 0 to 15cm with millimeter increments on one edge and 0 to 6 in. divided into 0.0625 in. units on the other edge
  • Serves as a measure of nucleic acid mobility in gels
Specifications
Specifications
Ruler, Fluorescent
Plastic