Learn More
Bel-Art™ SP Scienceware™ Glove Box Economy Sleeved Gloves
Compatible with Scienceware Portable Glove Box or any glove box with 6 in. glove ports
Brand:  Bel-Art™ H50029-0000
339.48 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 12398617
  • 409.11 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Durable neoprene fabrication
  • Safeguards against gas, liquid, or solid substance penetrants
  • Use with Clamping Ring, Part No. 03-421-102
  • Large, size 10
  • 1 pair/Pk.