missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Microtube Beta Shield
Prevents beta radiation.
Brand: Bel-Art™ F24980-0001
This item is not returnable.
View return policy
Description
- Solid clear-acrylic block with 13 wells (11mm dia.)
- Wells hold 1.5mL micro sample tubes in individual cells
- With tight-fitting cover allows space for micro tube flip tops
- Blocks Beta radiation including P-32, S-35 and C-14
- Shield ensures safe sample transport and saves bench space
Specifications
Acrylic | |
Clear Acrylic block with 13 wells | |
5.2 x 10.5 x 6.2 cm (2.0625 x 4.125 x 2.4375 po.) |
Safe sample transport | |
Micro Tube Beta Shield |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction