Learn More
Bel-Art™ SP Scienceware™ Mouse Restrainer with Dorsal Access
Quick, gentle immobilization allows hands-free dorsal access to perform subcutaneous injections without risk of being bitten and less likelihood of sticking fingers
Brand:  Bel-Art™ F46401-0000
Product Code. 12972590
  • 77.65 € / Each
Explore more special offers
Description
Description
  • Made from TPX™ and can be cleaned and sterilized with ethanol or autoclaved
  • Designed for use with mice weighing 18 to 35g
  • Easy-to-use entry funnel and two gate positions allows quick, gentle and secure immobilization of the mouse; both hands are now free to perform tasks
  • Mouse can be led head first or guided tail first into the restrainer. When used tail first, the tail remains outside the restrainer allowing for tail tattoos or tail bleeds
  • Circular opening in top of tube provides access to dorsal area allowing quick, accurate and consistent injections from mouse to mouse
  • Safe for use with ethanol and other common laboratory disinfectants; will not craze and weaken like acrylic restrainers
  • Autoclave-safe, permitting use with immunocompromised mice and other specially developed transgenic mice
  • Four built-in U-shaped channels on the base enables restrainer to be fastened to a surface to hold in place (hardware not included)
Specifications
Specifications
Autoclavable
Perform Subcutaneous Injections
2 gate positions, dorsal access
6.8 x 3.9 x 2.6 in. (17.4 x 10 x 6.7 cm)
17.4 cm
10 cm
6.7 cm
3.17 cm
Clear
Mouse Restrainer
TPX™ PMP
6.8 in.
3.9 in.
2.6 in.
1.25 in.
18 to 35 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Mouse Restrainer with Dorsal Access > Mouse Restrainer with Dorsal Access