missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Piezoelectric Gas Lighter
Needs no recharging
Brand: Bel-Art™ H15056-0000
67.30 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
Needs no recharging
- Compact, self-contained electric generator lights Bunsen burners, gas stoves, ovens, propane torches, and oxyacetylene torches
- Just hold in gas stream and press button
- Functions without flints, fuel, batteries, or wires
- Comfortable plastic pistol grip provides excellent control in aiming aluminum barrel
Order Info
Dimensions: 10 3/8L x 1W x 5 in.H (26 x 2.5 x 13cm).
Specifications
Piezo Electric Gas Lighter | |
26.5 cm | |
Comfortable plastic pistol grip provides excellent control |
10.438 in. | |
Plastic |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido