missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polypropylene Policeman

Each round a rod has one end flattened and cut diagonally

Brand:  Bel-Art™ F37739-0001

Product Code. 10212853

  • 27.40 € / Pack of 6
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Diameter 4.7mm (0.188 in.)
  • Length 20.3cm (8 in.)
  • Steam autoclavable at 121°C (250°F) for reuse without fear of sample contamination

Specifications

Specifications

Autoclavable
0.189 in.
Flattened end at one side and diagonally cut at other end
0.47 cm
Polypropylene
White
Designed for aggressive scraping and spreading without scratching
8 in.
20.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.