missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Polypropylene Sterilizing Trays

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

Brand:  Bel Art™ H16262-0000

 View more versions of this product

Product Code. 11752555

  • 66.00 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.

  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Trays are steam autoclavable at 121°C (250°F)
  • Covers are sold separately

Order Info

Covers are sold separately.

Specifications

Specifications

13 L
White
48.3 cm
Sterilizing Tray
Fits 13L Tray Cover, Mfr. No. H162610000
5 in.
10.5 in.
Autoclavable
19 x 10.5 x 5 in. (48.3 x 26.7 x 12.7 cm)
12.7 cm
Polypropylene
26.7 cm
19 in.
Rectangular
3.4 gal.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel Art™ Polypropylene Sterilizing Trays > Tray, 13L, 19 x 10.5 x 5 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.