Learn More
Bel-Art™ SP Scienceware™ Poxygrid™ Baskets
Epoxy-coated steel wire construction holds up under severe laboratory conditions
Brand:  Bel-Art™ H16759-0000
 View more versions of this product
Product Code. 11763207
  • 147.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Epoxy-coated steel wire construction holds up under severe laboratory conditions
  • Resist rust, corrosion, solvents, acids and alkalis
  • Convenient for transporting and batch-dipping glassware
  • Steam autoclavable at 121°C (250°F)
Specifications
Specifications
Epoxy-coated Steel
63 cm
36 cm
15.2 cm
24.75 x 14 x 6 in. (63 x 36 x 15.2cm)
24.75 in.
14 in.
6 in.