missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Poxygrid™ Petri Dish Dispensing Racks

Holds 11 to 33 plastic petri dishes. Bel Art™ SP Scienceware™ Poxygrid™ Petri Dish Dispensing Racks are durable and dispense dishes one at a time.

Brand:  Bel-Art™ F18978-0001

 View more versions of this product

Product Code. 11352194

  • 56.02 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Holds 11 to 33 plastic petri dishes. Bel Art™ SP Scienceware™ Poxygrid™ Petri Dish Dispensing Racks are durable and dispense dishes one at a time.

  • Durable epoxy-coated, steel wire construction
  • Dishes loaded at top and dispensed at bottom
  • Each channel holds up to eleven 100 x 20mm plastic Petri dishes
  • Tilts toward rear for convenient presentation of dishes
  • Not suitable for glass Petri dishes

Specifications

Specifications

Not Autoclavable
11
5.75 x 4.25 x 11 in. (14.6 x 10.7 x 28cm)
5.75 in.
4.25 in.
11 in.
Steel Wire
10 x 2cm Petri dishes
14.6 cm
10.7 cm
28 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.