Learn More
Bel-Art™ SP Scienceware™ Protective Labeling Tape System: Tape Dispenser
Utilizes a tape dispenser to fully cover and protect labels
Brand:  Bel-Art™ F13450-0000
Product Code. 10342671
  • 94.73 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Protective Labeling System provides labels that stand up to harsh laboratory conditions. The clear tape protects labels from the freezer, autoclave, acids, alkalis and organic solvents. Test results prove the tape remains securely affixed with no discoloration after being autoclaved for 40 minutes at 125°C (258°F), as well as after one week in liquid nitrogen at -196°C (-321°F). All tape sizes are 1mil (0.001 in.) thick with 1mil (0.001 in.) of acrylic adhesive.
  • Durable tape dispenser is molded of strong ABS plastic with a steel serrated cutting blade; tape roll held in place by aluminum roller and spring-loaded cover
  • Optional epoxy-coated, weighted steel base reduces slippage on the benchtop when tape is dispensed; includes four nonslip rubber feet and screws
  • Clear, polypropylene tape has high resistance to chemicals and moisture; provides great versatility for heavy-duty lab use
  • White paper labels accept any kind of marker; perfect for use under the clear tape to label labware, bottles or samples
Specifications
Specifications
ABS
Use to label containers, cabinets, drawers, books and lab equipment
1 Ea.
20.65 cm
2.38 in.
Tape Dispenser
Tape dispenser, Steel cutting blade and Aluminum roller
8.13 in.
4.62 in.
6 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products