Learn More
Bel-Art™ SP Scienceware™ Replacement Stopcocks
Polypropylene body has Teflon™ PTFE plug, washer and bright red handle
Brand:  Bel-Art™ F42040-0000
Product Code. 15109034
  • 61.50 € / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Polypropylene body has Teflon™ PTFE plug, washer and bright red handle
  • Three-way plug for vacuum draw, shut-off and vacuum release
  • Taper fits on desiccator porte
Use on Desiccator Models: 08-594-15A, 08-594-16A, 08-594-15B, 08-594-16B, 08-594-15C, 08-594-16C, 01-817-50
Specifications
Specifications
3-way Stopcock
Scienceware™ desiccators, space saver and Techni-Dome™ 360 models
PTFE
Polypropylene
Washer, high visibility bright red handle with nut
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Teflon™ is a registered trademark of the Chemours Company FC LLC used under license by Bel-Arts Products, Inc.