Learn More
Bel-Art™ SP Scienceware™ Riteflow™ Flowmeter Leveling Base Kit
Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube
Brand:  Bel-Art™ H40404-1000
Product Code. 10743694
  • 177.44 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 15.2cm (6 in.) Plexiglas™ triangular base
  • Three adjustable feet
  • Vial level
  • For use with Riteflow mounted flowmeters
Specifications
Specifications
Scienceware Flowmeters Catalog Nos.: 11-163-75 and -77 Series
Leveling Base Kit
Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube