Learn More
Bel-Art™ SP Scienceware™ Secador™ 1.0 Carrying Case Desiccators
For dry storage of moisture and UV sensitive products
Brand:  Bel-Art™ F42070-0001
 View more versions of this product
Product Code. 10710763
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Durastar™ co-polyester construction blocks 99% of UV light and is resistant to staining, crazing and chemical attack
  • Large door maximizes access to interior space and has a patented seal design, tab latches and security loops for a padlock or tamper-evident seals; opens top to bottom
  • Includes built-in suitcase style carrying handle and two removable shelves
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view dial hygrometer located on pull down oven style door
  • Can be used with desiccant cartridges F42048-0065 or F42046-0000
  • 16.3D × 8.4W × 14.7 in.H (41.4 × 21.3 × 37.3cm)
  • Internal volume: 0.75 cu. ft.
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626
Specifications
Specifications
Copolymer
For dry storage of moisture and UV sensitive products
14.7 in.
Secador™ Carrying Case, Blue
0.75 cu. ft
37.3 cm