Learn More
Bel-Art™ SP Scienceware™ Slide Tray Cabinet
Compact cabinet to organize and store microscope slides
Brand:  Bel-Art™ H44583-0000
Product Code. 10530754
  • 363.00 € / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Small footprint cabinet can be placed on the benchtop, making slides easily accessible
  • Dust-free storage
  • Holds 240 slides in 12 microscope trays (Mfr. No. H44581-0000, not included)
  • Polypropylene outer shell; polystyrene tray rails
  • Transparent vertically sliding acrylic door with door knob
  • 35.6L x 20.8W x 25.4cmH (14 x 83/16 x 10 in.)
Specifications
Specifications
240
356 mm
10 in.
Polypropylene, Polystyrene
131 mm
14 in.
240
254 mm
5.18 in.