missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Retriever

Reaches down drains for lost bars

Brand:  Bel-Art™ 08555-40

Product Code. 11705229

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Reaches down drains for lost bars

  • 0.38 in. (9.5mm) dia. polyethylene pick-up rod
  • With powerful Alnico magnet sealed in one end
  • Handy hang-up grip on other end
  • Offers convenient stir bar retrieval for bars up to 1 in. long

Specifications

Specifications

Offers convenient stir bar retrieval for bars up to 1 in. long
High-strength alnico sealed-in magnet, hang-up loop on one end, 0.95 cm polyethylene pick-up rod
45.7 cm
Spinbar™ Magnetic Retriever
Stirring bars
18 in.
Polyethylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.