Learn More
Bel-Art™ SP Scienceware™ Spinbox™ Spinplus™ Magnetic Stirring Bar Assortment
Useful assortments of popular sizes and shapes
Brand:  Bel-Art™ F37187-0000
 View more versions of this product
Product Code. 13203539
  • 248.00 € / Pack of 13
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Includes:
Octagon figured 16 assorted bars, standard and micro; standard bars with pivot ring Octagon shaped 12 assorted bars, standard bars with pivot rings Round shaped 22 assorted bars with removable pivot rings on bars longer than 1 in. Metric Polygon shaped 18 assorted bars Color-coded, octagon shaped 24 assorted bars with pivot ring Color-coded micro bars 12 assorted bars Egg shaped bars 13 assorted bars Spinplus in. + shaped bars 5 assorted bars
Description
Description
  • Convenient storage box segregates sizes and styles for easy, no-rummage organization
  • FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Available in octagon, egg and Spinplus™ shapes
  • Autoclavable
Certifications
ISO 9001:2008
Specifications
Specifications
Autoclavable
16 assorted standard and micro octagonal bars; 12 assorted standard octagonal bars with pivot ring; 22 assorted standard round bars with pivot ring (removable pivot ring on bars longer than 1 in.); 18 assorted metric polygon colour-coded bars; 24 assorted colour-coded micro bars with pivot ring; 12 assorted egg shaped bars; 13 assorted Spinplus including 5 '+' shaped bars
Egg
FDA, USP Class VI, ISO 9001:2008, SGS
Alnico Magnet