Learn More
Bel-Art™ Vaccu-Pette 96-Well Format Pipetting Device
Aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing or dispensing
Brand: Bel-Art™ F37876-0000
Alert:
For single use only: Cannot be autoclaved or gas sterilized
Description
Aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing or dispensing
- For simultaneously changing media or addition/removal of reagents from 96-well plate
- Connect disposable plastic syringe and withdraw plunger to transfer liquid
- Made of transparent plastic material
- With vacuum outlet at one end, and short tube of 3.2mm I.D. supplied for syringe connection
- Sterile/Disposable model molded of clear, disposable plastic
- Requires 30mL or larger disposable sterile syringe
Specifications
96 wells; L x W x H: 12.7 x 8.6 x 3.7cm | |
Sterile | |
Clear | |
12.7 cm | |
Reusable |
Plastic | |
Each | |
30 mL | |
Pipetting Device |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.