Learn More
Bel-Art™ Sterileware Economy Sampling Scoops
An inexpensive and accurate measure for dry or liquid sample collection.
Brand:  Bel-Art™ F36933-0002
 View more versions of this product
Product Code. 11332174
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
An inexpensive and accurate measure for dry or liquid sample collection.
  • Capacity is listed on each white scoop
  • Leveling the sample to the flat top edge ensures a repeatable and consistent volume measure
  • Flat bottom shape and light weight allows for use as a weighing boat
  • One-piece scoops are made from durable, FDA grade, high-impact polystyrene plastic
  • Sterile are individually sealed and sterilized in a polyethylene bag ready for one time use and disposal, lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
Scoop
Certificate of processing for sterilization
2.7 oz.
Single-use
White
Provide accurate measure for dry or liquid measures
50
1⅛ in.
2.9 cm
5⁹⁄₁₆ in.
1/2 in.
10.3 cm
1¹⁵⁄₁₆ in.
4.9 cm
High-impact Polystyrene (FDA Grade)
Flat
83 mL
Sterile
FDA
Sealed in polyethylene bag
14.1 x 4.9 x 2.9 cm (5⁹⁄₁₆ x 1¹⁵⁄₁₆ x 1⅛ in.)
1⅛ in.
2.9 cm
4¹⁄₁₆ in.
14.1 cm
3.8 cm
1¹⁵⁄₁₆ in.
4.9 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Sterilized products are not returnable.