missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Sterileware™ Sense-Able Scoops™ Metal and X-Ray Detectable Scoops

Scoop and scoop fragments can be detected by standard metal detection and x-ray equipment to prevent potential production contamination, quarantine and recall

Brand:  Bel-Art™ H369040005

 View more versions of this product

Product Code. 11725234

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Standard metal detection and x-ray equipment can detect scoop fragments as small as 1.6mm3
  • Molded with a custom FDA grade polystyrene resin blend containing trace amounts of stainless steel Dark blue or black color provides a high contrast background to better view material; Blue coloring conforms to industry standard for metal detectable materials
  • Long handles on 2 and 4 oz. scoops keeps hand farther from sample to reduce contamination or exposure
  • Flat bottom on 2 and 4 oz. scoops holds scoop upright for use a weighing boat: won't spill when resting on flat surface
  • Individually wrapped in polyethylene bag and gamma sterilized, lot stamped—ready for one-time use and disposal

Specifications

Specifications

Metal and X-Ray Detectable Scoop
Certificate of processing for sterilization
Long
125 mL
Sterile
Impregnated with Stainless Steel Powder
Prevents potential production contamination, quarantine and recal
1.31 in.
3.68 in.
9.4 cm
2 in.
Polystyrene Resin
Flat
4 oz.
Single-use
Black
FDA
Metal detection and X-ray equipment
3.3 cm
5 in.
12.7 cm
5.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.