Learn More
Bel-Art™ Super Biohazard Disposal Bags
Protects against dangerous tears and leaks. Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
Brand:  Bel-Art™ F13165-3138
 View more versions of this product
Product Code. 17682085
  • 1020.00 € / Pack of 200
Explore more special offers
Description
Description
  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization
Specifications
Specifications
25 gal.
31 x 38 in. (79 x 96 cm)
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
2 mil
Biohazard Disposal Bag
Red
Autoclavable
Extra-strength, high-impact, high-temperature bags reduce autoclave time
Polymer