Learn More
Bel-Art™ SP Scienceware™ Test Tube Capper
Eliminates fatigue and soreness of hands and fingers from the repetitive task of applying test tube caps
Brand: Bel-Art™ F18799-0000
This item is not returnable.
View return policy
Description
- Fits comfortably in the palm of the user's hand
- Tip applies pressure to securely cap tubes
- Capping can be performed with test tubes in a rack or while being held in the hand
- Designed for use with recessed center test tube caps
- Polypropylene with vinyl-covered handhold
Specifications
Test Tube Capper | |
Polypropylene | |
Eliminates fatigue and soreness of hands and fingers from the repetitive task of applying test tube caps |