missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Transpette™ Eight Channel Transfer Pipettor

For washing plates, feeding, and supernatant aspiration from microwell plates

67.00 € - 393.15 €

Specifications

For Use With (Application) For washing plates, feeding, and supernatant aspiration from microwell plates
Color Natural
Volume (Metric) 600 μL
Length (Metric) 180 mm
Type Transfer Pipet
View More Specs

Products 2
Product Code Brand Sterility Material Packaging Type Price Quantity & Availability  
Product Code Brand Sterility Material Packaging Type Price Quantity & Availability  
10075210
View Documents
Bel-Art™
F37873-0001
Sterile Polyethylene Individually Wrapped
393.15 €
Pack of 100
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
11844160
View Documents
Bel-Art™
F37873-0000
Non-sterile Polypropylene Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

For washing plates, feeding, and supernatant aspiration from microwell plates

  • 8-Channel with 600μL capacity per channel
  • Made of translucent plastic
  • Discard after hot radioactive uses to avoid radiation contamination of expensive liquid handling devices
  • Non-autoclavable
  • Available with sterile and non-sterile options
Specifications

Specifications

For washing plates, feeding, and supernatant aspiration from microwell plates
600 μL
Transfer Pipet
Natural
180 mm
Single-use
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Transpette™ Eight Channel Transfer Pipettor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.