Learn More
Bel-Art™ Vaccu-Pette 96-Well Format Pipetting Device, Autoclavable
Aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing, dispensing or aspirating of wells by working simultaneously.
Brand: Bel-Art™ F37876-0001
53.14 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Requires : 30mL or larger disposable sterile syringe
This item is not returnable. View return policy
Description
Aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing, dispensing or aspirating of wells by working simultaneously.
- For simultaneously changing media or the addition/removal of reagents from a 96 well plate
- Simply connect a disposable plastic syringe and withdraw the plunger to transfer liquid
- Made of transparent plastic material, it has vacuum outlet at one end, and a short length of 3.2mm I.D. tubing is supplied for syringe connection
- Autoclavable/Reusable model is molded of polycarbonate; must be sterilized by autoclaving at no more than 121°C for 20 minutes at 15 psi prior to use in order to avoid contamination
- Cannot be gas sterilized because of the possibility of residual trapped gases
- Requires a 30mL or larger disposable sterile syringe (not included)
Specifications
No | |
Clear | |
96-well Plates | |
Polycarbonate | |
96 | |
No | |
Pipetting Device | |
12.7 x 8.6 x 3.7cm (5 x 3.38 x 1.44 in.) |
Autoclavable | |
Reusable | |
Ergonomic | |
Vaccu-pette | |
SP Scienceware Vaccu-Pette | |
Non-sterile | |
30 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.