missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Vakuwash™ Spectrophotometer Cell Washer

Cuvet washing device cleans without damaging

Brand:  Bel-Art™ F38960-0000

205.93 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 11396954

  • 221.00 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket

Includes: 500mL polypropylene filtering flask, Vikem™ stopper and PVC tube to connect to vacuum source.
This item is not returnable. View return policy

Description

Description

Cuvet washing device cleans without damaging

  • Simple to use cleaner connects to any vacuum source
  • Partially fill funnel with cleaning solution and press open end of cell (or tube or vial) against soft rubber seal to force cleaning solution to spray into cell
  • Used fluids collect in flask for reuse
  • Continued pressure against cell creates airflow for drying
  • Use acetone as final wash before drying to help remove all water droplets
  • Minimum cuvet, tube or vial O.D. must be 9mm to form seal over the spray nozzle

Specifications

Specifications

Plastic
500 mL polypropylene vacuum flask, Vikem™ vinyl stopper and PVC tube to connect to vacuum source
50 Hz
Glassware Washer
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ Vakuwash™ Spectrophotometer Cell Washer > Vakuwash cell and Cuvet Washer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.