missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wire Lead Flask with Vikem™ Coating

Wire lead weights coil into a helix, spiral, cone or any irregular shape to stabilize vessels in a water bath

Brand:  Bel-Art™ F18324-0000

 View more versions of this product

Product Code. 11397174

  • 39.21 € / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Can be coiled around flasks, threaded through handles or looped around bottle necks
  • Coated with chemical resistant red Vikem vinyl
  • The 30cm (12 in.) length and flexibility make these weights very versatile

Specifications

Specifications

Accessory
0.25 in.
0.25 lb.
Not Autoclavable
0.64 cm
114 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.