missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ ProCulture™ Micro-Tube Homogenizer: Pestles and Tubes

Product Code. 17142486 Shop All SP Scienceware Products
Click to view available options
Packaging Type:
Bulk Pack
Individual Pack
Sample Volume (Metric):
0.5 mL
1.5 mL
Product Type:
Pestle
Pestle and Tube
Tube
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 17142486

Brand: Bel-Art™ F650000004

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical and heat resistant, colorant-free

Matched pestle tips and micro tubes can be used either with homogenizer or with hand processing.

  • Tight tolerances, thorough homogenization and grinding of materials
  • Chemical and heat resistant, colorant-free to eliminate potential leaching of heavy metals into samples
  • Manufactured and packaged in class 10,000 cleanroom
  • Certified DNase, RNase and Pyrogen-free
  • Made of animal and phalate/phalate-ester free polypropylene
  • Available in individual or bulk packs
Pestles
  • Unique ribbed shaft provides precise finger control of homogenization for shear sensitive samples
  • Fit most motorized mixer adapters
Tubes
  • Tube bottom 25% thicker bottoms than industry standard
  • Frosted side and cap surfaces for labeling, positive seal snap-cap design and pierceable top

Specifications

Product Type Tube
Voltage 0/1 V
Sample Volume (Metric) 1.5 mL
Material Polypropylene
Dnase Free Yes
For Use With (Equipment) Micro-Tube homogenizer system
Packaging Quantity 100/Pk.
Packaging Type Individual Pack
RNase Free Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.