missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Bench-Top Mini-Bottle Roller Apparatus

For cell cultivation research or staining procedures. Wheaton™ Bench-Top Mini-Bottle Roller Apparatus allows for serum and media bottles, as well as culture tubes.

Brand:  Wheaton™ W348923-D

2565.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12412082

  • 2770.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accommodates bottles 38 to 60mm in diameter and 240mm long
  • Deck kits available to double or triple capacity
  • Precise and reproducible speed adjustments
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 to 45 rpm
100/240 V
14 W
50/60 Hz
18 cm
8 kg
For the growth and observation of various cell cultures
100/240 V, 50/60 Hz, 14 W
40°C
CE, CSA, UL
32.5 cm
32 cm
D Model
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers