missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Bench-top Stand for Waste Disposal Bags

This Bench-top Stand for Waste Disposal Bags stands 35 cm high and accommodates waste disposal bags sized 300 x 500 mm.

Brand:  Ratiolab™ 7001430

Product Code. 16201462

  • 22.05 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Practical bench-top stand, plastic-coated, with 3 rubber feet.
  • For bag size 300 x 500 mm

waste, disposable, stand, bag

Specifications

Specifications

Waste Disposal Bags, 300 x 500 mm
Non-sterile
35 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.