Learn More
Koettermann™ Heavy-Duty Table, SGL Laboratory Worktop
Heavy-duty mobile table suitable for a variety of laboratory tasks. Koettermann™ Heavy-Duty Table, SGL Laboratory Worktop is designed for durability and resistance.
Brand: Koettermann™ 507-00027
Description
- Suitable for a variety of tasks:
- Writing
- Chemical and equipment handling
- Hygiene and aseptic requirement areas
- Surdy and durable:
- Sturdy castors
- Resistant to chemicals
- Scratch resistant
- Maximum loading capacity: 400kg
Specifications
Mobile Table | |
900 mm | |
150 cm | |
1 |
1500 x 750 x 900 mm | |
Trespa TopLab Worktop | |
400 kg | |
750 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.